Merck
  • Associative and dissociative mechanisms in the formation of phthalazine bridged organodiplatinum(II) complexes.
Associative and dissociative mechanisms in the formation of phthalazine bridged organodiplatinum(II) complexes.
Inorganic chemistry (2010-08-18)
Mehdi Rashidi, S Masoud Nabavizadeh, Ahad Zare, Sirous Jamali, Richard J Puddephatt
ZUSAMMENFASSUNG
The reaction of phthalazine with the binuclear organoplatinum complexes [Me(2)Pt(μ-SMe(2))(μ-dppm)PtR(2)], R = Me, Ph, 4-tolyl or R(2) = (CH(2))(4), dppm = bis(diphenylphosphino)methane, gives the corresponding complexes [Me(2)Pt(μ-phthalazine)(μ-dppm)PtR(2)] by displacement of the bridging dimethylsulfide ligand. The structures of [Me(2)Pt(μ-SMe(2))(μ-dppm)PtMe(2)] and [Me(2)Pt(μ-phthalazine)(μ-dppm)PtMe(2)] have been determined. Kinetic studies show that the reactions occur mostly by a second order reaction when R = Me or R(2) = (CH(2))(4) but entirely by a first order reaction when R = Ph or 4-tolyl. Evidence is presented that the reactions when R = Me or R(2) = (CH(2))(4) can occur by either associative or dissociative mechanisms but that the reactions when R = Ph or 4-tolyl occur only by an unusual dissociative mechanism involving formation of an intermediate with a donor-acceptor Pt-Pt bond.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phthalazin, 98%