Merck
  • Regulation of lithospermic acid B and shikonin production in Lithospermum erythrorhizon cell suspension cultures.
Regulation of lithospermic acid B and shikonin production in Lithospermum erythrorhizon cell suspension cultures.
Chemical & pharmaceutical bulletin (2002-08-23)
Hirobumi Yamamoto, Ping Zhao, Kazufumi Yazaki, Kenichiro Inoue
ZUSAMMENFASSUNG
Cell suspension cultures of Lithospermum erythrorhizon produced a large amount of lithospermic acid B, a caffeic acid tetramer, as well as shikonin derivatives (each ca. 10% of dry wt.) when cultured in shikonin production medium M-9. Various culture factors for increasing the production of lithospermic acid B were investigated. Lithospermic acid B production was inhibited by 2, 4-D or NH4+, whereas it was stimulated by Cu2+. These regulatory patterns were similar to those for the production of shikonin derivatives in these cell cultures, suggestive of close relations and similar metabolic regulation between the production of these compounds. Cultivation under light illumination, however, showed that these metabolisms were independently regulated. In particular, blue light showed a stimulatory effect on lithospermic acid B production, while shikonin production was strongly inhibited, indicative of an effective condition for lithospermic acid B production.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salvianolsäure B, ≥94% (HPLC)
Supelco
Salvianolsäure B, analytical standard