Merck
  • Sequence of the cDNA encoding human GP-2, the major membrane protein in the secretory granule of the exocrine pancreas.
Sequence of the cDNA encoding human GP-2, the major membrane protein in the secretory granule of the exocrine pancreas.
Gene (1996-06-01)
S M Wong, A W Lowe
ZUSAMMENFASSUNG
GP-2 is the major membrane protein in the pancreatic secretory granule of the exocrine pancreas. The nucleotide sequence of the human GP-2 gene was determined from cDNA clones isolated from a lambda gt11 pancreatic library.