Merck
  • Ferrocenyl chalcones versus organic chalcones: a comparative study of their nematocidal activity.
Ferrocenyl chalcones versus organic chalcones: a comparative study of their nematocidal activity.
Bioorganic & medicinal chemistry (2011-02-26)
Saeed Attar, Zachary O'Brien, Hasan Alhaddad, Melissa L Golden, Alejandro Calderón-Urrea
ZUSAMMENFASSUNG
A series of 30 organic chlacones and 33 ferrocenyl (Fc) chalcones were synthesized and characterized by melting point, elemental analysis, spectroscopy ((1)H NMR and FTIR) and, in two cases, by X-ray crystallography. The biological activity of each compound (10(-4)M in DMSO) against the model nematode Caenorhabditis elegans was examined in terms of % mortality (percent nematodes that died) and % fecundity (percent nematodes that reproduced) and compared to that obtained for the control medium (1% DMSO) over a 14-day period. Detailed conformational analyses for two Fc-chalcones (studied also by X-ray crystallography) were performed via molecular modeling studies. In general, the organic chalcones were found to be less polar than their Fc analogs. Some structure-activity relationships (SARs) were determined: (a) The nematocidal activities of the organic chalcones in this series were found to be much greater than those of their ferrocenyl analogs. (b) The position of the carbonyl group played a central role in the biological activity of both classes of chalcones studied. (c) For both classes of chalcones, lipophilicity of a compound seemed to play a significant role in its nematocidal activity. (d) The planarity of a ferrocenyl-chlacone seems to play a role in its activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
trans-Chalkon, 97%