Merck
  • Capacitive sensor based on molecularly imprinted polymers for detection of the insecticide imidacloprid in water.
Capacitive sensor based on molecularly imprinted polymers for detection of the insecticide imidacloprid in water.
Scientific reports (2020-09-04)
Suzan El-Akaad, Mona A Mohamed, Nada S Abdelwahab, Eglal A Abdelaleem, Sarah De Saeger, Natalia Beloglazova
ZUSAMMENFASSUNG
This manuscript reports on the development of a capacitive sensor for the detection of imidacloprid (IMD) in water samples based on molecularly imprinted polymers (MIPs). MIPs used as recognition elements were synthesized via a photo-initiated emulsion polymerization. The particles were carefully washed using a methanol (MeOH) /acetic acid mixture to ensure complete template removal and were then dried. The average size of the obtained particles was less than 1 µm. The imprinting factor (IF) for IMD was 6 and the selectivity factor (α) for acetamiprid, clothianidin, thiacloprid and thiamethoxam were 14.8, 6.8, 7.1 and 8.2, respectively. The particles were immobilized on the surface of a gold electrode by electropolymerization. The immobilized electrode could be spontaneously regenerated using a mixture of MeOH/10 mM of phosphate buffer (pH = 7.2)/triethylamine before each measurement and could be reused for 32 times. This is the first-time that automated regeneration was introduced as part of a sensing platform for IMD detection. The developed sensor was validated by the analysis of artificially spiked water samples. Under the optimal conditions, the linearity was in the range of 5-100 µM, with a limit of detection (LOD) of 4.61 µM.