Merck
Abundance and localization of human UBE3A protein isoforms.
Human molecular genetics (2020-08-25)
Carissa L Sirois, Judy E Bloom, James J Fink, Dea Gorka, Steffen Keller, Noelle D Germain, Eric S Levine, Stormy J Chamberlain
ZUSAMMENFASSUNG
Loss of UBE3A expression, a gene regulated by genomic imprinting, causes Angelman syndrome (AS), a rare neurodevelopmental disorder. The UBE3A gene encodes an E3 ubiquitin ligase with three known protein isoforms in humans. Studies in mouse suggest that the human isoforms may have differences in localization and neuronal function. A recent case study reported mild AS phenotypes in individuals lacking one specific isoform. Here we have used CRISPR/Cas9 to generate isogenic human embryonic stem cells (hESCs) that lack the individual protein isoforms. We demonstrate that isoform 1 accounts for the majority of UBE3A protein in hESCs and neurons. We also show that UBE3A predominantly localizes to the cytoplasm in both wild type and isoform-null cells. Finally, we show that neurons lacking isoform 1 display a less severe electrophysiological AS phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-E6AP antibody, Mouse monoclonal, clone E6AP-330, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-UBE3A antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution