Merck
Improved Performance of Ternary Solar Cells by Using BODIPY Triads.
Materials (Basel, Switzerland) (2020-06-19)
Sompit Wanwong, Weradesh Sangkhun, Pisist Kumnorkaew, Jatuphorn Wootthikanokkhan
ZUSAMMENFASSUNG
Two boron dipyrromethene (BODIPY) triads, namely BODIPY-1 and BODIPY-2, were synthesized and incorporated with poly-3-hexyl thiophene: (6,6)-phenyl-C61-butyric acid methyl ester (PCBM) P3HT:PCBM. The photovoltaic performance of BODIPY:P3HT:PCBM ternary solar cells was increased, as compared to the control binary solar cells (P3HT:PCBM). The optimized power conversion efficiency (PCE) of BODIPY-1:P3HT:PCBM was improved from 2.22% to 3.43%. The enhancement of PCE was attributed to cascade charge transfer, an improved external quantum efficiency (EQE) with increased short circuit current (Jsc), and more homogeneous morphology in the ternary blend.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(Diphenylamino)phenylboronsäure, ≥95%