Merck
  • Clusterin contributes to hepatitis C virus-related hepatocellular carcinoma by regulating autophagy.
Clusterin contributes to hepatitis C virus-related hepatocellular carcinoma by regulating autophagy.
Life sciences (2020-06-07)
Na Fu, Huijuan Du, Dongdong Li, Yu Lu, Wencong Li, Yang Wang, Lingbo Kong, Jinghua Du, Suxian Zhao, Weiguang Ren, Fang Han, Rongqi Wang, Yuguo Zhang, Yuemin Nan
ZUSAMMENFASSUNG
To explore the potential regulatory mechanism of differentially expressed mRNAs in Hepatitis C virus (HCV)-related hepatocellular carcinoma (HCC). Patients with HCV-related HCC and age- and gender-matched healthy subjects were enrolled. Differentially expressed mRNAs in the plasma were detected by digital gene expression (DGE) profile analysis. HepG2 and SMMC7721 cells stably transfected with HCV-core protein and the control plasmid were established. And small interfering RNA (siRNA) was used to knockdown the target gene in HCV core-expressing HCC cell lines. mRNA expression was determined by qRT-PCR. Protein expression was measured by Western blot and immunohistochemistry staining. DGE profile data showed aberrant mRNA expression contributed to the progression of HCV-HCC, and clusterin (CLU), which was significantly highly expressed, was chosen as a candidate gene. Further evidence showed CLU was highly expressed in tumor tissues of HCV-HCC patients and HCV core-expressing HCC cell lines, accompanied with enhanced autophagy and upregulation of pro-autophagy genes. And knockdown of CLU in HCC cell lines suppressed cell autophagy, which was indicated by decreased expression of autophagy marker light chain 3B (LC3B) ІІ/І ratio, and downregulated pro-autophagy genes like Beclin1, autophagy-related protein 7 (Atg7) and Lamp2. On the other hand, anti-autophagy genes or regulators, including p62 and phosphorylated mammalian target of rapamycin (p-mTOR), were notably upregulated. CLU could promote the progression of HCV-related HCC by regulating autophagy, which might be a potential therapeutic target of HCV-HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human CLU