Merck
Facile access to nitroarenes and nitroheteroarenes using N-nitrosaccharin.
Nature communications (2019-08-01)
Roxan Calvo, Kun Zhang, Alessandro Passera, Dmitry Katayev
ZUSAMMENFASSUNG
Nitroaromatics and nitroheteroaromatics serve as key building blocks and intermediates in synthesis, and form the core scaffold of a vast number of materials, dyes, explosives, agrochemicals and pharmaceuticals. However, their synthesis relies on harsh methodologies involving excess mineral acids, which present a number of critical drawbacks in terms of functional group compatibility and environmental impact. Modern, alternative strategies still suffer from significant limitations in terms of practicality, and a general protocol amenable to the direct C-H functionalization of a broad range of aromatics has remained elusive. Herein we introduce a bench-stable, inexpensive, easy to synthesize and recyclable nitrating reagent based on saccharin. This reagent acts as a controllable source of the nitronium ion, allowing mild and practical nitration of both arenes and heteroarenes displaying an exceptional functional group tolerance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Nitrosaccharin