Merck
  • Arterial Wall Stress Induces Phenotypic Switching of Arterial Smooth Muscle Cells in Vascular Remodeling by Activating the YAP/TAZ Signaling Pathway.
Arterial Wall Stress Induces Phenotypic Switching of Arterial Smooth Muscle Cells in Vascular Remodeling by Activating the YAP/TAZ Signaling Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-11-23)
Yongshun Wang, Wei Cao, Jinjin Cui, Yang Yu, Yubo Zhao, Jian Shi, Jian Wu, Zhengyuan Xia, Bo Yu, Jingjin Liu
ZUSAMMENFASSUNG
Increasing wall stress or biomechanical stretch experienced by arteries influences the initiation of atherosclerotic lesions. This initiation is mediated by Yes-associated protein (YAP) and transcriptional co-activator with PDZ-binding motif (TAZ), which are both effectors of the Hippo pathway. In this study, the functional roles of YAP/TAZ proteins in the regulation of the stretch-mediated programing of human umbilical arterial smooth muscle cells (HUASMCs) to a proliferative phenotype were examined. HUASMCs were seeded on a Matrigel-coated silicone chamber and subjected to biomechanical stretch for 24 h after 48 h of growth. YAP/TAZ small interfering RNA was used to specifically knockdown YAP/ TAZ expression in HUASMCs. We observed that YAP/TAZ activation via biomechanical stretching is involved in the regulation of critical aspects of the HUASMC phenotypic switch. YAP/TAZ knockdown significantly attenuated the stretch-induced proliferative and pro-inflammatory phenotypes in HUASMCs. Furthermore, treatment with atorvastatin, an anti-atherosclerotic drug, attenuated the stretch-induced phenotypic switch of HUASMCs from the contractile to synthetic state by suppressing YAP/TAZ expression. Additional investigations demonstrated the role of stretch in inhibiting the Hippo pathway, leading to the activation of PI3-kinase (PI3K) and phosphoinositide dependent kinase (PDK1); the key molecule for the regulation of the PDK1 and Hippo complex interaction was Sav1. These results showed the importance of YAP/TAZ activation, induced by biomechanical stretch, in promoting atheroprone phenotypes in HUASMCs. Taken together, our findings revealed a mechanism by which YAP/TAZ activation contributes to the pathogenesis of atherosclerosis.