Merck
  • Amperometric aptasensor for carcinoembryonic antigen based on the use of bifunctionalized Janus nanoparticles as biorecognition-signaling element.
Amperometric aptasensor for carcinoembryonic antigen based on the use of bifunctionalized Janus nanoparticles as biorecognition-signaling element.
Analytica chimica acta (2019-03-31)
Gonzalo Paniagua, Anabel Villalonga, Marcos Eguílaz, Borja Vegas, Concepción Parrado, Gustavo Rivas, Paula Díez, Reynaldo Villalonga
ZUSAMMENFASSUNG
We report herein the design of a novel biosensing strategy for the detection of carcinoembryonic antigen (CEA), based on the use of Janus-type nanoparticles having Au and silica opposite faces as integrated electrochemical biorecognition-signaling system. The Janus nanoparticles were properly functionalized with horseradish peroxidase on the silica surface to act as signaling element, and a biotin thiol-modified anti-CEA DNA hairpin aptamer the Au face to assemble the biorecognition element. The sensing approach relies on the first specific recognition of CEA by the bifunctionalized Janus nanoparticles, causing unfolding of the DNA hairpin structure and unmasking the biotin residues at the aptamer chain. This CEA-Janus nanoparticle complex was then captured by avidin-modified Fe3O4@SiO2 NanoCaptors®, allowing further magnetic deposition on carbon screen printed electrodes for the amperometric detection of the cancer biomarker. The Janus nanoparticles-based aptasensor was able to detect CEA in the range from 1 to 5000 ng mL-1 (5.5 pM-28 nM) with a detection limit of 210 pg mL-1 (1.2 pM). The aptasensor also showed high reproducibility and storage stability, and was successfully validated in human serum.