Merck
Black carp IRF5 interacts with TBK1 to trigger cell death following viral infection.
Developmental and comparative immunology (2019-07-02)
Can Yang, Liqun Liu, Ji Liu, Zi Ye, Hui Wu, Pinghui Feng, Hao Feng
ZUSAMMENFASSUNG
Interferon regulated factor 5 (IRF5) is a key regulator of inflammatory responses in human and mammals; however, its role in teleost remains largely unknown. In this study, IRF5 homologue of black carp (Mylopharyngodon piceus) has been cloned and characterized, which possesses conservation in structure and sequence to its mammalian counterparts. Black carp IRF5 (bcIRF5) was characterized as a predominantly cytosolic protein by immunofluorescent staining and showed little IFN promoter-inducing ability in reporter assay. The direct association between bcIRF5 and black carp TBK1 (bcTBK1) were identified through co-immunoprecipitation assay, and co-expressed bcIRF5 in EPC cells suppressed bcTBK1-mediated IFN promoter transcription in reporter assay. Surprisingly, the titer of grass carp reovirus (GCRV) in the media of EPC cells co-expressing bcIRF5 and bcTBK1 was obviously lower than that of EPC cells expressing bcTBK1 alone. It was interesting that expression of bcIRF5 and/or bcTBK1 in EPC cells showed little effect on cell growth; however, the survival ratio of EPC cells co-expressing bcTBK1 and bcIRF5 post GCRV infection was much lower than that of EPC cells expressing bcIRF5 or bcTBK1 alone. These results indicate that bcIRF5 negatively regulates bcTBK1-mediated IFN signaling in healthy cells; however, it correlates with bcTBK1 and triggers cell death to inhibit the virus replication during the innate immune activation.