Merck
S100A9 extends lifespan in insulin deficiency.
Nature communications (2019-08-09)
Giorgio Ramadori, Sanda Ljubicic, Serena Ricci, Despoina Mikropoulou, Xavier Brenachot, Christelle Veyrat-Durebex, Ebru Aras, Rafael M Ioris, Jordi Altirriba, Elisabeth Malle, Dirk Foell, Thomas Vogl, Roberto Coppari
ZUSAMMENFASSUNG
Tens of millions suffer from insulin deficiency (ID); a defect leading to severe metabolic imbalance and death. The only means for management of ID is insulin therapy; yet, this approach is sub-optimal and causes life-threatening hypoglycemia. Hence, ID represents a great medical and societal challenge. Here we report that S100A9, also known as Calgranulin B or Myeloid-Related Protein 14 (MRP14), is a leptin-induced circulating cue exerting beneficial anti-diabetic action. In murine models of ID, enhanced expression of S100A9 alone (i.e. without administered insulin and/or leptin) slightly improves hyperglycemia, and normalizes key metabolic defects (e.g. hyperketonemia, hypertriglyceridemia, and increased hepatic fatty acid oxidation; FAO), and extends lifespan by at least a factor of two. Mechanistically, we report that Toll-Like Receptor 4 (TLR4) is required, at least in part, for the metabolic-improving and pro-survival effects of S100A9. Thus, our data identify the S100A9/TLR4 axis as a putative target for ID care.