- alpha-thiocyanation of carbonyl and beta-dicarbonyl compounds using (dichloroiodo)benzene-lead(II) thiocyanate.
alpha-thiocyanation of carbonyl and beta-dicarbonyl compounds using (dichloroiodo)benzene-lead(II) thiocyanate.
The Journal of organic chemistry (2001-04-13)
O Prakash, H Kaur, H Batra, N Rani, S P Singh, R M Moriarty
PMID11300895
ZUSAMMENFASSUNG
The combination reagent (dichloroiodo)benzene and lead(II) thiocyanate in dichloromethane effects oxidation of various enol silyl ethers, ketene silyl acetals, and beta-dicarbonyl compounds, thereby providing an efficient and convenient method for alpha-thiocyanation of carbonyl and beta-dicarbonyl compounds.