Merck
Specific inhibitors of vacuolar type H(+)-ATPases induce apoptotic cell death.
Biochemical and biophysical research communications (1995-07-06)
T Nishihara, S Akifusa, T Koseki, S Kato, M Muro, N Hanada
ZUSAMMENFASSUNG
Concanamycin A and bafilomycin A1 are known as strong inhibitors of the vacuolar type H(+)-ATPases in vitro. These inhibitors exhibited cytotoxic effects on twelve cell lines in cell viability assay. On the other hand, the F1F0-type H(+)-ATPase inhibitor oligomycin and the E1E2-type H(+)-ATPase inhibitor vanadate showed no cytotoxic effect. We show here that concanamycin A and bafilomycin A1 induce a significant increase in the proportion of fragmented DNA in agarose gel electrophoresis. Flow cytometric cell cycle analysis of WEHI 231 cells stimulated with concanamycin A revealed the increased percentage of apoptotic cells with hypodiploid DNA. These findings indicate that cell death induced by specific inhibitors of vacuolar type H(+)-ATPases occurs through apoptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Concanamycin A, ≥70% (HPLC)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Bafilomycin A1, InSolution, ≥90%, A macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge