Merck
HomeProtein ExpressionPhosphoprotein Phosphatases (Serine/Threonine)
Phosphoprotein Phosphatases (Serine/Threonine)
Phosphoprotein phosphatases, which hydrolyze the phosphoester bonds of phosphoserines, phosphothreonines or phosphotyrosines, play an essential role in signal transduction and actively contribute to the regulation of protein phosphorylation. On the basis of their substrate specificity they are usually divided into phosphoserine and phosphothreonine phosphatases on the one hand, and phosphotyrosine and dual-specificity phosphatases, on the other hand. This division corresponds also to different families of enzymes with different catalytic mechanisms. Genes coding for phosphoserine/threonine phosphatases are less numerous in vertebrate genomes than those for serine /threonine kinases, and the complexity of phosphatases function arises in part from the interactions of catalytic subunits with other proteins.
Prior to the knowledge of their sequence, phosphoserine/threonine phosphatases were classified on the basis of their substrate preference and inhibitor sensitivity. Type 1 protein phosphatases (PP1) dephosphorylate the β-subunit of phosphorylase kinase and are inhibited by phospho-inhibitor-1 and inhibitor-2, whereas type 2 protein phosphatases (PP2) dephosphorylate the α-subunit of phosphorylase kinase and are resistant to the aforementioned inhibitors. Type 2 phosphatases were further divided depending on their divalent cation requirements for full activity: phosphatase 2A (PP2A) does not require such cations, whereas phosphatase 2B (PP2B, also known as calcineurin) and phosphatase 2C (PP2C) are respectively activated by Ca2+ and Mg2+.
Molecular cloning revealed that serine/threonine phosphatases belong to two different families of about a dozen of genes each in mammals: the phosphoprotein phosphatase P (PPP) family includes PP1, PP2A, PP2B, and a few related enzymes, PP4, PP5, PP6 and PP7, while the phosphoprotein phosphatase M family (PPM) includes PP2C and related enzymes. Sequences of the catalytic domains of PPP family phosphatases are highly related to one another. They have been highly conserved during evolution and are homologous in general aspects of their catalytic mechanism to enzymes found in prokaryotes (e.g. phage l phosphatase). PPM phosphatases are found in prokaryotes and eukaryotes, and are numerous in plants. Although PPP and PPM families share no sequence homology, they are both metallo-enzymes and their 3-D structures display some general similarities.
The serine/threonine phosphatases of the PPP family are mostly regulated by protein-protein interactions. Both PP1c and PP2Ac are catalytic subunits that interact with a large number of targeting and regulatory subunits. PP1 is targeted to glycogen particles, myofibrils or dendritic spines by specific subunits and regulated by small proteins controlled by phosphorylation. For example, in basal ganglia neurons, PP1c is inhibited by the phosphorylated form of DARPP-32 (32 kDa dopamine and cAMP-regulated protein) that is regulated by dopamine and several other neurotransmitters. The PP2A catalytic subunit forms heterotrimers with two other subunits and interacts with several inhibitory proteins. In contrast, PP2B (calcineurin) is comprised of an A subunit including the catalytic domain and a B subunit structurally related to calmodulin. Calcineurin is activated by direct Ca2+ binding to calcineurin B and binding of the Ca2+-calmodulin complex to calcineurin A. PP7 is also activated by Ca2+.
A number of natural toxins produced by a variety of microorganisms are powerful inhibitors of PP1, PP2A, and PP4-6, with only relatively small differences in affinities. This, combined with the high molar concentrations of these phosphatases found in some cells, makes it sometimes difficult to identify their relative contribution to the dephosphorylation of specific substrates in intact cells through the use of these inhibitors. Calcineurin is the specific target of two immunosuppressant drugs, cyclosporin A and FK506, complexed with their intracellular receptors (immunophilins), cyclophilin and FK506-binding protein (FKBP), respectively.
PPM family members are monomeric enzymes for which regulatory mechanisms or targeting by protein-protein interactions are as yet poorly characterized. It includes the classic PP2C phosphatases, a number of enzymes recently discovered and pyruvate dehydrogenase phosphatases. There are as yet no specific inhibitor of PPM phosphatases.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Footnotes
a) Binding protein, some inhibitory effect.
b) Inhibitors are listed in order of decreasing potency.
c) Combined with cyclophilin.
d) Combined with FKBP.
Abbreviations
AKAP-79: A kinase anchor protein 79
Cain: Calcineurin inhibitor
CaM: Calmodulin
CPI17: 17 kDa PKC-potentiated inhibitory protein of PPI
DARPP32: Dopamine and cAMP-regulated phosphoprotein
DSCR1: Down syndrome critical region 1
FKBP: FK506 binding protein
I1PP2A: Protein inhibitor 1 of protein phosphatase 2A
I2PP2A: Protein inhibitor 2 of protein phosphatase 2A
Inhibitor-2: Proteon phosphatase inhibitor-2
NIPP: Nuclear inhibitor of protein phosphatase 1
PP: Phosphoprotein phosphatase
PP1: Phosphoprotein phosphatase 1
PP2A: Phosphoprotein phosphatase 2A
PP2B: Phosphoprotein phosphatase 2B or calcineurin
PP2C: Phosphoprotein phosphatase 2C
PPM: Phosphoprotein phosphatase M family
PPP: Phosphoprotein phosphatase P family
RIPP: Ribosomal inhibitor of protein phosphatase 1
ZAKI-4: Down syndrome critical region 1-like 1
Similar Products
Loading
References
1.
Barford D, Das AK, Egloff M. 1998. THE STRUCTURE AND MECHANISM OF PROTEIN PHOSPHATASES: Insights into Catalysis and Regulation. Annu. Rev. Biophys. Biomol. Struct.. 27(1):133-164. https://doi.org/10.1146/annurev.biophys.27.1.133
2.
Butler T, Paul J, Europe-Finner N, Smith R, Chan E. 2013. Role of serine-threonine phosphoprotein phosphatases in smooth muscle contractility. American Journal of Physiology-Cell Physiology. 304(6):C485-C504. https://doi.org/10.1152/ajpcell.00161.2012
3.
Campbell BE, Hofmann A, McCluskey A, Gasser RB. 2011. Serine/threonine phosphatases in socioeconomically important parasitic nematodes?Prospects as novel drug targets?. Biotechnology Advances. 29(1):28-39. https://doi.org/10.1016/j.biotechadv.2010.08.008
4.
Cohen P. 1991. [33] Classification of protein- serine/threonine phosphatases: identification and quantitation in cell extracts.389-398. https://doi.org/10.1016/0076-6879(91)01035-z
5.
Cohen PT. 1997. Novel protein serine/threonine phosphatases: Variety is the spice of life. Trends in Biochemical Sciences. 22(7):245-251. https://doi.org/10.1016/s0968-0004(97)01060-8
6.
Forrest AR. 2003. Phosphoregulators: Protein Kinases and Protein Phosphatases of Mouse. Genome Research. 13(6):1443-1454. https://doi.org/10.1101/gr.954803
7.
Gehringer MM. 2004. Microcystin-LR and okadaic acid-induced cellular effects: a dualistic response. 557(1-3):1-8. https://doi.org/10.1016/s0014-5793(03)01447-9
8.
Honkanen R, Golden T. 2002. Regulators of Serine / Threonine Protein Phosphatases at the Dawn of a Clinical Era?. CMC. 9(22):2055-2075. https://doi.org/10.2174/0929867023368836
9.
KENNELLY PJ. 2003. Archaeal protein kinases and protein phosphatases: insights from genomics and biochemistry. 370(2):373-389. https://doi.org/10.1042/bj20021547
10.
Millward TA, Zolnierowicz S, Hemmings BA. 1999. Regulation of protein kinase cascades by protein phosphatase 2A. Trends in Biochemical Sciences. 24(5):186-191. https://doi.org/10.1016/s0968-0004(99)01375-4
11.
Munton RP, Vizi S, Mansuy IM. 2004. The role of protein phosphatase-1 in the modulation of synaptic and structural plasticity. 567(1):121-128. https://doi.org/10.1016/j.febslet.2004.03.121
12.
Peng A, Maller JL. 2010. Serine/threonine phosphatases in the DNA damage response and cancer. Oncogene. 29(45):5977-5988. https://doi.org/10.1038/onc.2010.371
13.
Pereira SR, Vasconcelos VM, Antunes A. 2011. The phosphoprotein phosphatase family of Ser/Thr phosphatases as principal targets of naturally occurring toxins. Critical Reviews in Toxicology. 41(2):83-110. https://doi.org/10.3109/10408444.2010.515564
14.
Schweighofer A, Hirt H, Meskiene I. 2004. Plant PP2C phosphatases: emerging functions in stress signaling. Trends in Plant Science. 9(5):236-243. https://doi.org/10.1016/j.tplants.2004.03.007
15.
Svenningsson P, Nishi A, Fisone G, Girault J, Nairn AC, Greengard P. 2004. DARPP-32: An Integrator of Neurotransmission. Annu. Rev. Pharmacol. Toxicol.. 44(1):269-296. https://doi.org/10.1146/annurev.pharmtox.44.101802.121415
16.
Wera S, Hemmings BA. 1995. Serine/threonine protein phosphatases. 311(1):17-29. https://doi.org/10.1042/bj3110017
17.
Wurzenberger C, Gerlich DW. 2011. Phosphatases: providing safe passage through mitotic exit. Nat Rev Mol Cell Biol. 12(8):469-482. https://doi.org/10.1038/nrm3149