Merck
HomeNanoparticle & Microparticle SynthesisPerovskite Quantum Dots Synthesis
Perovskite Quantum Dots Synthesis
Qingchao Zhou, Feng Zhang, Haizheng Zhong
School of Materials Science & Engineering, Beijing Institute of Technology, Beijing, 100081, China
Introduction
Colloidal quantum dots (QDs) are solution-processable luminescent materials that can produce high-quality color in light emitting diodes (LEDs). These LEDs are essential components of advanced solid-state lighting and flat panel displays.1 After more than 30 years of development, CdSe and InP based QDs are now emerging as “upstarts” in commercial liquid crystal display (LCD) TVs from TCL (XESS X1) and Samsung (SUHD), as well as prototype EL display demos from BOE (SID 2017). However, large-scale commercialization is still limited by high cost and limited reliability. This performance gap has helped to generate intense interest in finding new approaches. QD-LEDs are based on either a down conversion photoluminescence (PL) mechanism or on an electricity-light conversion electroluminescence (EL) mechanism.2,3 QDs (or nanocrystals) comprised of halide perovskites (ABX3, where A = Cs+ or CH3NH3+ (methyl ammonium; MA) or CH2(NH2)2+ (formamidinium; FA) and X = Br, Cl or I) QDs or nanocrystals show outstanding characteristics including superior PL properties, low fabrication cost, and enhanced color performance.4-6 These features make them promising candidates as light emitters for display technology.7-10
As illustrated in Figure 1, the development of perovskite QDs dates back to the discovery of enhanced PL emissions in nanosized perovskite embedded in meso-Al2O3 in 2012.11 Publications on this topic have shown exponential growth after highly luminescent colloidal perovskite QDs were first reported in 2015.9,10,12 Progress has been made in the fabrication of perovskite QDs as strategies have been developed to fabricate high-quality materials, enabling rapid light emitting device development. Even when compared to well-developed CdSe and InP QDs, prototype color conversion LEDs based on perovskite QDs show superior color performance and improved luminous efficiency (120% NTSC, 109 lm/W), enabling enhanced LCD panels (Figure 2A).13 Meanwhile, the performance of perovskite QD-based EL devices now approaches that of other conventional QDs, even showing promise for use in flexible displays (Figure 2B).14 The controllable fabrication of high quality perovskite QDs is a prerequisite for application exploration. In this mini-review, we highlight some of the most highly-developed synthetic techniques for perovskite QDs, as well as provide a brief summary of the progress toward display applications.
Publications with respect to perovskite QDs
Figure 1.Publications with respect to perovskite QDs. Compiled using Web of Science database and title = (perovskite quantum dots) or title = (perovskite nanocrystal).
Images of the flexible perovskite QD-based LEDs and devices tailored with abbreviations of “UCLA & BIT.”
Figure 2.  A) Images of the flexible perovskite QD-based LEDs and devices tailored with abbreviations of “UCLA & BIT.”   B) Comparison of perovskite QDs enhanced LCD screen (left) and MacBook LCD screen (right).
Hot Injection Method
The hot injection technique was the first method introduced for the colloidal synthesis of CdSe QDs, and it has since been widely applied to a variety of colloidal nanoparticles.15,16 By adapting this technique, colloidal perovskite nanocrystals can be fabricated in a supersaturated solution through fast injection of the precursors at an elevated temperature.9,17-19 In 2014, Schmidt et al. first reported the preparation of colloidal CH3NH3PbBr3 nanoparticles by mixing CH3(CH2)17NH3Br/CH3NH3Br with PbBr2 at 80 °C.12 However, the resulting CH3NH3PbBr3 nanoparticles exhibited only moderate PL quantum yield (PLQY) of ~20% and showed poor dispersity in colloidal solution. After optimization, however, the authors were able to achieve a significantly higher PLQY of 87%.20 Soon thereafter, Kovalenko et al. adapted the hot-injection technique to fabricate all inorganic CsPbX3 QDs with an even higher PLQY of ~90%.9 To accomplish this the authors used a PbX2 type material such as PbI2, PbBr2, PbCl2 or a mixture thereof, together with oleylamine (OLA) and oleic acid (OA, Product No. O1008), dissolved in 1-octadecene (ODE, Product No. 74740). An injection of Cs-oleate precursor at an elevated temperature of 140-200 °C initiated the nucleation and growth of highly luminescent colloidal CsPbX3 QDs with tunable emission spectra and PLQYs up to 90% (Figure 3B). However, CsPbX3 QDs synthesized by the hot injection method exhibited poor colloidal stability and rapidly precipitated from the crude solution. To solve this, Yassitepe et al. developed an improved amine-free strategy using OA as the surface ligand, obtaining enhanced colloidal stability and enhanced performance in EL devices.21 Protesescu et al. fabricated CH2(NH2)2 + doped CsPbI3 QDs by injecting a mixed precursor of CH2(NH2)2 + oleate and Cs-oleate.22 Other groups, including Rogach et al. explored other inorganic ligands, reporting the surface protection of CsPbX3 (X = Br or I) with a polyhedral oligomeric silsesquioxane.23
A schematic diagram of hot injection synthesis of perovskite QDs/NCs
Figure 3.  A) Schematic diagram of hot injection synthesis of perovskite QDs/NCs.   B) Perovskite QDs/NCs in toluene under UV lamp (λ = 365 nm) and corresponding PL spectra.9 Reproduced with permission: copyright 2015 American Chemical Society.
Ligand Assisted Reprecipitation (LARP)
Reprecipitation through solvent mixing is a simple way to prepare organic nanoparticles or polymer dots simultaneously.25-27 Hybrid perovskites are soluble in many polar solvents, but are insoluble in non-polar ones. Based on the miscible pair of toluene and N,N-dimethylformamide (DMF, Product No. D158550900638, etc.), our group demonstrated ligand-assisted reprecipitation synthesis (LARP) to fabricate perovskite QDs via solvent mixing with the assistance of long chain organic ligands.10 Figure 4A schematically illustrates the LARP technique. In a typical synthesis of CH3NH3PbX3 QDs, the precursor solution (PbX2, CH3NH3Br, n-octylamine, and OA) was dropped into toluene. The change in solubility subsequently induces the formation of highly luminescent colloidal QDs. The investigation of solvent-precursor interactions helped provide insight on the influence of solvents on the formation process and obtained stable iodide-based CH3NH3PbI3 quantum dots, as well as morphology-controlled perovskite nanocrystals.28,29
A schematic diagram of the ligand-assisted reprecipitation technique (LARP) for perovskite QDs/NCs
Figure 4.  A) Schematic diagram of the ligand-assisted reprecipitation technique (LARP) for perovskite QDs/NCs.   B) Perovskite QDs/NCs in toluene under UV lamp (λ = 365 nm) and corresponding PL spectra.10 Reproduced with permission: copyright 2015 American Chemical Society.
The LARP technique is both simple and versatile for the synthesis of halide perovskite QDs and other nanostructures. For example, Xiong et al. used it to synthesize amorphous pervoskite QDs and achieved bright EL devices.30 Jie et al. developed a simple dip-coating method to effectively fabricate large-area and uniformly dense films for multicolor EL devices.31 Other groups have adopted the LARP strategy for the synthesis of other perovskite QDs, including CsPbX3 QDs, CH2(NH2)2PbX3 and (CH3NH3)3Bi2X9 QDs.32-34 However, LARP still has some shortcomings as a synthetic route for device applications, such as low yields of CH3NH3PbX3 QDs, and the difficulty of cleaning and purifying the synthesized colloidal solutions, which greatly hinders the application of perovskite QDs to EL devices. Based on the immiscible solvents hexane and DMF, our group has further developed the emulsion synthesis method to obtain purified CH3NH3PbX3 QDs for use in EL device applications.35 Sun et al. reported using thermally-induced crosslinking or radical-induced polymerization to address the stability issue and produce more efficient EL devices.36 Further, many groups are working toward the application of down conversion white LEDs for LCD backlights by capping and sealing perovskites QDs during or after their formation. The inclusion of insulating barrier coatings such as SiO2 or POSS also has potential to overcome stability issues.23,37,38
Mechanochemical Methods
Both the hot injection technique and LARP synthesis utilize organic solvents and surface ligands, and both require difficult isolation and purification steps. Solid-state mechanosynthesis avoids the use of organic solvents, and has the potential to achieve scalable production.39,40 This strategy is depicted in Figure 5, where metal salts of AX and PbX2 are used as the immediate raw materials, producing reaction products in a single step after simple milling/grinding at ambient temperature.41 Highly luminescent QDs can then be obtained by further grinding/milling and surface treatment of the perovskite product with appropriate ligands such as OLA. Mechanical mixing, especially high-speed milling, induces heat and supplies reaction energy. Sonication is also effective at breaking bulk perovskite powder into nanoparticles.42
A schematic diagram of mechanochemical method (milling/grinding/sonication) for perovskite nanocrystals.
Figure 5.Schematic diagram of mechanochemical method (milling/grinding/sonication) for perovskite nanocrystals.
In situ Formation Strategy
For device applications, fabricated QDs must be processed into composite or thin films. However, forming composite films with homogenous distribution or uniform films on proper interfacial layers remains a significant technical challenge. Because perovskites can be processed at room temperature, there are many ways to simplify the process for display applications using an in situ formation strategy. For example, Dirin et al. investigated the growth of perovskite nanocrystals in mesoporous silica (meso-SiO2) matrixes,43 and showed that it can serve as a facile, complementary, and ligand-free preparation route for outstanding perovskite-based composites with broadly tunable emission from the green to near-infrared. In this method, shown in Figure 6, highly concentrated solutions of the precursor salts, AX and PbX2, were inserted into the pores (width 2.5-50 nm) of various commercially available mesoporous silica templates, followed by the removal of excess solution, and then by drying induced the spatial crystallization of perovskite nanocrystals.
The use of mesoporous silica templates allows for the inexpensive and scalable preparation of lead halide perovskite composites. However, the inorganic matrix serves as a hard template and therefore, similar to the traditional rare-earth phosphors, the as-fabricated perovskite based composites require further encapsulation. The hard template does not offer good protection against aggregation, and it has been reported that significant decomposition occurs during the annealing of perovskite NCs in porous TiO2 at 85 °C, even in an inert atmosphere.
Regarding LCD backlight applications, a more appealing approach is the in situ fabrication of perovskite QD based composite films (PQDCF). The polymer matrix tends to form a coherent barrier layer around the perovskite QDs/NCs, protecting them from water, oxygen, and heat of the surrounding environment. Using this method, our group has demonstrated the in situ fabrication of CH3NH3PbX3 QDs embedded in polyvinylidene fluoride (PVDF) composite films by separating the crystallization processes of PVDF and CH3NH3PbX3 QDs through controlled evaporation of DMF.13 As illustrated in Figure 6, a typical fabrication of CH3NH3PbBr3 based PQDCF involved two main stages. In stage I, a mixture of MABr, PbBr2 and PVDF was dissolved in DMF, and then a uniform precursor film was obtained through solvent evaporation. In stage II, the colorless precursor films were moved out to remove the residual DMF, gradually resulting in green emissive PQDCF. Brightly-luminescent and color-tunable PQDCF with emissions ranging from 440 nm to 730 nm could be similarly obtained by varying the compositions of halides. These PQDCFs show enhanced PL properties with high PLQYs up to 90% (±5%), and improved stability against both moisture and UV radiation, making them useful for on-surface LCD backlight applications. A prototype LED device showed excellent merits in both color quality and luminous efficiency, optimal for display backlights. Soon after, Wang et al. reported a swellingdeswelling microencapsulation strategy that achieved welldispersed, intimately passivated perovskite QDs inside polymer matrixes that had high photoluminescence efficiency, color purity and ultrahigh stability against both heat and water.44
Among these several methods, the in situ formation strategy is currently the most promising method for achieving practical display applications, due to its simplified, low-cost, and largescale processing, as well as the enhanced photoluminescence and stability of the product.
A schematic diagram of in situ formation strategy for perovskite QDs/NCs within inorganic and polymer matrices.
Figure 6.Schematic diagram of in situ formation strategy for perovskite QDs/NCs within inorganic and polymer matrices.
Device Perspective
By adapting the synthetic strategies as previously described, perovskite QDs with superior PL properties have been successfully fabricated and explored for use in display applications. Although studies on perovskite QDs are still in their infancy, tremendous progress has been made toward overcoming the material challenges and technical problems of conventional QDs. By adapting the available techniques from well-developed CdSe QDs, the fundamental framework for implementation in display system appears to be well established. Figure 7 describes the important features of CdSe, InP, and perovskite QDs. For LCD backlighting, perovskite QDs are advantageous in brightness and color gamut, but suffer from stability issues. Because of their easy scale-up and simple processing, mechanosynthesis and in situ formation are preferred for the fabrication of perovskite-based phosphors for LCD backlights. Building on these advances, the continuous work on material stability and device optimization has already generated the first TV demonstration at CES 2018 by TCL (International Consumer Electronics Show), which is expected to be commercialized in the coming year.
Apart from display applications, the perovskite QDs have also been investigated for solar cell applications. Recent work shows that under operation, perovskite solar cells using these materials retained 90% (97% after dark recovery) of their initial performance after 500 hours of continuous room temperature operation at their maximum power point under 1-sun illumination, a promising development.45 Although the stability of perovskite QDs is still under debate, there is no doubt that perovskite QDs are highly promising candidates for EL devices. Currently, the device performance of perovskite QD-based EL devices (QLED) still lag behind welldeveloped CdSe QDs. Solution processing methods such as hot injection and LARP are expected to fabricate purified QDs that can be used in flexible printing QLED displays. We are looking forward to continued improvements in device performance, as well as growing applications for perovskite QDs in other display technologies such as light emitting cells and laser displays.
A comparison of the important featured of different ads for display applications
Figure 7.Comparison of the important featured of different ads for display applications.
Materials
Loading
References
1.
Shirasaki Y, Supran GJ, Bawendi MG, Bulovi? V. 2013. Emergence of colloidal quantum-dot light-emitting technologies. Nature Photon. 7(1):13-23. https://doi.org/10.1038/nphoton.2012.328
3.
Jang E, Jun S, Jang H, Lim J, Kim B, Kim Y. White-Light-Emitting Diodes with Quantum Dot Color Converters for Display Backlights. Adv. Mater.. 22(28):3076-3080. https://doi.org/10.1002/adma.201000525
4.
Bai Z, Zhong H. 2015. Halide perovskite quantum dots: potential candidates for display technology. Science Bulletin. 60(18):1622-1624. https://doi.org/10.1007/s11434-015-0884-y
5.
Yang G, Zhong H. 2016. Organometal halide perovskite quantum dots: synthesis, optical properties, and display applications. Chinese Chemical Letters. 27(8):1124-1130. https://doi.org/10.1016/j.cclet.2016.06.047
6.
Huang H, Polavarapu L, Sichert JA, Susha AS, Urban AS, Rogach AL. 2016. Colloidal lead halide perovskite nanocrystals: synthesis, optical properties and applications. NPG Asia Mater. 8(11):e328-e328. https://doi.org/10.1038/am.2016.167
7.
Cho H, Jeong S, Park M, Kim Y, Wolf C, Lee C, Heo JH, Sadhanala A, Myoung N, Yoo S, et al. 2015. Overcoming the electroluminescence efficiency limitations of perovskite light-emitting diodes. Science. 350(6265):1222-1225. https://doi.org/10.1126/science.aad1818
8.
Stranks SD, Snaith HJ. 2015. Metal-halide perovskites for photovoltaic and light-emitting devices. Nature Nanotech. 10(5):391-402. https://doi.org/10.1038/nnano.2015.90
9.
Protesescu L, Yakunin S, Bodnarchuk MI, Krieg F, Caputo R, Hendon CH, Yang RX, Walsh A, Kovalenko MV. 2015. Nanocrystals of Cesium Lead Halide Perovskites (CsPbX3, X = Cl, Br, and I): Novel Optoelectronic Materials Showing Bright Emission with Wide Color Gamut. Nano Lett.. 15(6):3692-3696. https://doi.org/10.1021/nl5048779
10.
Zhang F, Zhong H, Chen C, Wu X, Hu X, Huang H, Han J, Zou B, Dong Y. 2015. Brightly Luminescent and Color-Tunable Colloidal CH3NH3PbX3 (X = Br, I, Cl) Quantum Dots: Potential Alternatives for Display Technology. ACS Nano. 9(4):4533-4542. https://doi.org/10.1021/acsnano.5b01154
11.
Kojima A, Ikegami M, Teshima K, Miyasaka T. 2012. Highly Luminescent Lead Bromide Perovskite Nanoparticles Synthesized with Porous Alumina Media. Chem. Lett.. 41(4):397-399. https://doi.org/10.1246/cl.2012.397
12.
Schmidt LC, Pertegás A, González-Carrero S, Malinkiewicz O, Agouram S, Mínguez Espallargas G, Bolink HJ, Galian RE, Pérez-Prieto J. 2014. Nontemplate Synthesis of CH3NH3PbBr3 Perovskite Nanoparticles. J. Am. Chem. Soc.. 136(3):850-853. https://doi.org/10.1021/ja4109209
13.
Zhou Q, Bai Z, Lu W, Wang Y, Zou B, Zhong H. 2016. In Situ Fabrication of Halide Perovskite Nanocrystal-Embedded Polymer Composite Films with Enhanced Photoluminescence for Display Backlights. Adv. Mater.. 28(41):9163-9168. https://doi.org/10.1002/adma.201602651
14.
Zhao F, Chen D, Chang S, Huang H, Tong K, Xiao C, Chou S, Zhong H, Pei Q. Highly flexible organometal halide perovskite quantum dot based light-emitting diodes on a silver nanowire?polymer composite electrode. J. Mater. Chem. C. 5(3):531-538. https://doi.org/10.1039/c6tc04934f
15.
Murray CB, Norris DJ, Bawendi MG. 1993. Synthesis and characterization of nearly monodisperse CdE (E = sulfur, selenium, tellurium) semiconductor nanocrystallites. J. Am. Chem. Soc.. 115(19):8706-8715. https://doi.org/10.1021/ja00072a025
16.
Zhong H, Mirkovic T, Scholes G. 2011. Nanocrystal Synthesis.153-201. https://doi.org/10.1016/b978-0-12-374396-1.00051-9
17.
Pan A, He B, Fan X, Liu Z, Urban JJ, Alivisatos AP, He L, Liu Y. 2016. Insight into the Ligand-Mediated Synthesis of Colloidal CsPbBr3 Perovskite Nanocrystals: The Role of Organic Acid, Base, and Cesium Precursors. ACS Nano. 10(8):7943-7954. https://doi.org/10.1021/acsnano.6b03863
18.
Song J, Li J, Li X, Xu L, Dong Y, Zeng H. 2015. Quantum Dot Light-Emitting Diodes Based on Inorganic Perovskite Cesium Lead Halides (CsPbX3). Adv. Mater.. 27(44):7162-7167. https://doi.org/10.1002/adma.201502567
19.
Hassan Y, Song Y, Pensack RD, Abdelrahman AI, Kobayashi Y, Winnik MA, Scholes GD. 2016. Structure-Tuned Lead Halide Perovskite Nanocrystals. Adv. Mater.. 28(3):566-573. https://doi.org/10.1002/adma.201503461
20.
Gonzalez-Carrero S, Galian RE, Pérez-Prieto J. Maximizing the emissive properties of CH3NH3PbBr3 perovskite nanoparticles. J. Mater. Chem. A. 3(17):9187-9193. https://doi.org/10.1039/c4ta05878j
21.
Yassitepe E, Yang Z, Voznyy O, Kim Y, Walters G, Castañeda JA, Kanjanaboos P, Yuan M, Gong X, Fan F, et al. 2016. Amine-Free Synthesis of Cesium Lead Halide Perovskite Quantum Dots for Efficient Light-Emitting Diodes. Adv. Funct. Mater.. 26(47):8757-8763. https://doi.org/10.1002/adfm.201604580
22.
Protesescu L, Yakunin S, Kumar S, Bär J, Bertolotti F, Masciocchi N, Guagliardi A, Grotevent M, Shorubalko I, Bodnarchuk MI, et al. 2017. Dismantling the ?Red Wall? of Colloidal Perovskites: Highly Luminescent Formamidinium and Formamidinium?Cesium Lead Iodide Nanocrystals. ACS Nano. 11(3):3119-3134. https://doi.org/10.1021/acsnano.7b00116
23.
Huang H, Chen B, Wang Z, Hung TF, Susha AS, Zhong H, Rogach AL. Water resistant CsPbX3 nanocrystals coated with polyhedral oligomeric silsesquioxane and their use as solid state luminophores in all-perovskite white light-emitting devices. Chem. Sci.. 7(9):5699-5703. https://doi.org/10.1039/c6sc01758d
24.
Zhang D, Eaton SW, Yu Y, Dou L, Yang P. 2015. Solution-Phase Synthesis of Cesium Lead Halide Perovskite Nanowires. J. Am. Chem. Soc.. 137(29):9230-9233. https://doi.org/10.1021/jacs.5b05404
25.
Kasai H, Nalwa HS, Oikawa H, Okada S, Matsuda H, Minami N, Kakuta A, Ono K, Mukoh A, Nakanishi H. 1992. A Novel Preparation Method of Organic Microcrystals. Jpn. J. Appl. Phys.. 31(Part 2, No. 8A):L1132-L1134. https://doi.org/10.1143/jjap.31.l1132
26.
Zhao YS, Fu H, Peng A, Ma Y, Xiao D, Yao J. 2008. Low-Dimensional Nanomaterials Based on Small Organic Molecules: Preparation and Optoelectronic Properties. Adv. Mater.. 20(15):2859-2876. https://doi.org/10.1002/adma.200800604
27.
Niu Y, Zhang F, Bai Z, Dong Y, Yang J, Liu R, Zou B, Li J, Zhong H. 2015. Aggregation-Induced Emission Features of Organometal Halide Perovskites and Their Fluorescence Probe Applications. Advanced Optical Materials. 3(1):112-119. https://doi.org/10.1002/adom.201400403
28.
Zhang F, Chen C, Kershaw SV, Xiao C, Han J, Zou B, Wu X, Chang S, Dong Y, Rogach AL, et al. 2017. Ligand-Controlled Formation and Photoluminescence Properties of CH3NH3PbBr3 Nanocubes and Nanowires. ChemNanoMat. 3(5):303-310. https://doi.org/10.1002/cnma.201700034
29.
Zhang F, Huang S, Wang P, Chen X, Zhao S, Dong Y, Zhong H. 2017. Colloidal Synthesis of Air-Stable CH3NH3PbI3 Quantum Dots by Gaining Chemical Insight into the Solvent Effects. Chem. Mater.. 29(8):3793-3799. https://doi.org/10.1021/acs.chemmater.7b01100
30.
Xing J, Yan F, Zhao Y, Chen S, Yu H, Zhang Q, Zeng R, Demir HV, Sun X, Huan A, et al. 2016. High-Efficiency Light-Emitting Diodes of Organometal Halide Perovskite Amorphous Nanoparticles. ACS Nano. 10(7):6623-6630. https://doi.org/10.1021/acsnano.6b01540
31.
Deng W, Xu X, Zhang X, Zhang Y, Jin X, Wang L, Lee S, Jie J. 2016. Organometal Halide Perovskite Quantum Dot Light-Emitting Diodes. Adv. Funct. Mater.. 26(26):4797-4802. https://doi.org/10.1002/adfm.201601054
32.
Li X, Wu Y, Zhang S, Cai B, Gu Y, Song J, Zeng H. 2016. CsPbX3Quantum Dots for Lighting and Displays: Room-Temperature Synthesis, Photoluminescence Superiorities, Underlying Origins and White Light-Emitting Diodes. Adv. Funct. Mater.. 26(15):2435-2445. https://doi.org/10.1002/adfm.201600109
33.
Minh DN, Kim J, Hyon J, Sim JH, Sowlih HH, Seo C, Nam J, Eom S, Suk S, Lee S, et al. 2017. Room-Temperature Synthesis of Widely Tunable Formamidinium Lead Halide Perovskite Nanocrystals. Chem. Mater.. 29(13):5713-5719. https://doi.org/10.1021/acs.chemmater.7b01705
34.
Leng M, Chen Z, Yang Y, Li Z, Zeng K, Li K, Niu G, He Y, Zhou Q, Tang J. 2016. Lead-Free, Blue Emitting Bismuth Halide Perovskite Quantum Dots. Angew. Chem. Int. Ed.. 55(48):15012-15016. https://doi.org/10.1002/anie.201608160
35.
Huang H, Zhao F, Liu L, Zhang F, Wu X, Shi L, Zou B, Pei Q, Zhong H. 2015. Emulsion Synthesis of Size-Tunable CH3NH3PbBr3 Quantum Dots: An Alternative Route toward Efficient Light-Emitting Diodes. ACS Appl. Mater. Interfaces. 7(51):28128-28133. https://doi.org/10.1021/acsami.5b10373
36.
Sun H, Yang Z, Wei M, Sun W, Li X, Ye S, Zhao Y, Tan H, Kynaston EL, Schon TB, et al. 2017. Chemically Addressable Perovskite Nanocrystals for Light-Emitting Applications. Adv. Mater.. 29(34):1701153. https://doi.org/10.1002/adma.201701153
37.
Huang S, Li Z, Kong L, Zhu N, Shan A, Li L. 2016. Enhancing the Stability of CH3NH3PbBr3 Quantum Dots by Embedding in Silica Spheres Derived from Tetramethyl Orthosilicate in ?Waterless? Toluene. J. Am. Chem. Soc.. 138(18):5749-5752. https://doi.org/10.1021/jacs.5b13101
38.
Wang H, Lin S, Tang A, Singh BP, Tong H, Chen C, Lee Y, Tsai T, Liu R. 2016. Mesoporous Silica Particles Integrated with All-Inorganic CsPbBr3 Perovskite Quantum-Dot Nanocomposites (MP-PQDs) with High Stability and Wide Color Gamut Used for Backlight Display. Angew. Chem. Int. Ed.. 55(28):7924-7929. https://doi.org/10.1002/anie.201603698
39.
Prochowicz D, Franckevi?ius M, Cie?lak AM, Zakeeruddin SM, Grätzel M, Lewi?ski J. Mechanosynthesis of the hybrid perovskite CH3NH3PbI3: characterization and the corresponding solar cell efficiency. J. Mater. Chem. A. 3(41):20772-20777. https://doi.org/10.1039/c5ta04904k
40.
Jodlowski AD, Yépez A, Luque R, Camacho L, de?Miguel G. 2016. Benign-by-Design Solventless Mechanochemical Synthesis of Three-, Two-, and One-Dimensional Hybrid Perovskites. Angew. Chem. Int. Ed.. 55(48):14972-14977. https://doi.org/10.1002/anie.201607397
41.
Zhu Z, Yang Q, Gao L, Zhang L, Shi A, Sun C, Wang Q, Zhang H. 2017. Solvent-Free Mechanosynthesis of Composition-Tunable Cesium Lead Halide Perovskite Quantum Dots. J. Phys. Chem. Lett.. 8(7):1610-1614. https://doi.org/10.1021/acs.jpclett.7b00431
42.
Hintermayr VA, Richter AF, Ehrat F, Döblinger M, Vanderlinden W, Sichert JA, Tong Y, Polavarapu L, Feldmann J, Urban AS. 2016. Tuning the Optical Properties of Perovskite Nanoplatelets through Composition and Thickness by Ligand-Assisted Exfoliation. Adv. Mater.. 28(43):9478-9485. https://doi.org/10.1002/adma.201602897
43.
Dirin DN, Protesescu L, Trummer D, Kochetygov IV, Yakunin S, Krumeich F, Stadie NP, Kovalenko MV. 2016. Harnessing Defect-Tolerance at the Nanoscale: Highly Luminescent Lead Halide Perovskite Nanocrystals in Mesoporous Silica Matrixes. Nano Lett.. 16(9):5866-5874. https://doi.org/10.1021/acs.nanolett.6b02688
44.
Wang Y, He J, Chen H, Chen J, Zhu R, Ma P, Towers A, Lin Y, Gesquiere AJ, Wu S, et al. 2016. Ultrastable, Highly Luminescent Organic-Inorganic Perovskite-Polymer Composite Films. Adv. Mater.. 28(48):10710-10717. https://doi.org/10.1002/adma.201603964
45.
Tan H, Jain A, Voznyy O, Lan X, García de Arquer FP, Fan JZ, Quintero-Bermudez R, Yuan M, Zhang B, Zhao Y, et al. 2017. Efficient and stable solution-processed planar perovskite solar cells via contact passivation. Science. 355(6326):722-726. https://doi.org/10.1126/science.aai9081