HPLC Analysis of Adenosine Monophosphate (AMP) and Adenosine Triphosphate (ATP) on SeQuant® ZIC-HILIC
Materials
accessory
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),coated tip
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),coated tip
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),coated tip
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 20 x 2.1 mm, 3.5 μm, 100 Å (1.50439.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate
2.0 mL/min
pressure
1886 psi (13.1 MPa)
detector
254 nm
injection
0.75 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany