Merck
UHPLC Gradient Analysis of Boswellia Herbal Supplement on Ascentis® Express Phenyl-Hexyl
UHPLC Gradient Analysis of Boswellia Herbal Supplement on Ascentis® Express Phenyl-Hexyl application for UHPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Phenyl-Hexyl, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 3 mm
mobile phase component
Product No.
Description
Pricing
Phosphoric acid
≥85 wt. % in H2O, ≥99.999% trace metals basis
CONDITIONS
column
Ascentis Express Phenyl-Hexyl, 10 cm x 3.0 mm I.D., 2.7 μm particles (53345-U)
mobile phase
[A] 0.1% phosphoric acid; [B] acetonitrile
gradient
50% B for 0.5 min; to 100% B in 4.5 min; held at 100% B for 5 min
gradient
0.6 mL/min
pressure
2390 psi (165 bar)
column temp.
35 °C
detector
UV, 210 nm
injection
2 μL
Description
Application
extraction:
  1. Weigh 100 mg Bosewellia Herbal Supplement
    2. Add 10 mL ethanol
    3. Sonicate 30 min at 40°C
    4. Dilute 100 μL extract, 400 μL ethanol, 500 μL water
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany