Merck
HPLC Analysis of B-vitamins on Ascentis® Express RP-Amide using UV detection
HPLC Analysis of B-vitamins on Ascentis® Express RP-Amide using UV detection application for UHPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 3 cm × 4.6 mm
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 3.0 mm I.D., 2.7 μm particles (53918-U)
mobile phase
[A] 20 mM potassium phosphate, pH 3; [B] methanol;
gradient
0.5% B held for 1.5 minutes, to 25% B in 1.0 minutes, held at 25% B for 1.7 minute, held at 0.5% B for 1.2 minutes
flow rate
0.75 mL/min
pressure
5584 psi (385 bar)
column temp.
30 °C
detector
UV, 210 nm
injection
0.5 μL
sample
100 μg/mL of all vitamins except 250 μg/mL of calcium pantothenate, biotin and riboflavin in water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany