HPLC Analysis of Proteins and Peptides on Proteomix® WCX-NP1.7 Demonstrating Lot-to-Lot Reproducibility
![HPLC Analysis of Proteins and Peptides on Proteomix® WCX-NP1.7 Demonstrating Lot-to-Lot Reproducibility application for HPLC](/deepweb/assets/sigmaaldrich/product/images/211/800/c5c9102c-180b-4ce2-95a5-a933c00a5a77/800/c5c9102c-180b-4ce2-95a5-a933c00a5a77.jpg)
Materials
analytical column
Product No.
Description
Pricing
Proteomix® WCX-NP1.7 Weak Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 1.7 μm particle size, stainless steelCONDITIONS
column
Proteomix WCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 μm particles (Z777180)
mobile phase
(A) 20 mM sodium phosphate, pH 6.5; (B) A + 1.0 M NaCl
gradient
0 to 100% B in 25 min, 15 min prewash
flow rate
0.5 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
1 mg/mL
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies