HPLC Analysis of Nateglinide on Purospher® STAR RP-18 endcapped
CONDITIONS
column
Purospher STAR RP-18 endcapped 150 x 4.6 mm (1.51455.0001)
mobile phase
[A] 0.05% trifluoroacetic acid solution; [B] acetonitrile; (40:60, A:B)
flow rate
1 mL/min
pressure
1160 psi (80 bar)
column temp.
30 °C
detector
210 nm cell: 1 μL/10mm
injection
10 μL
sample
30 μg/mL (30 ppm) of Nateglinide in diluent to obtain the standard solution.
Description
Legal Information
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany