Merck
HPLC Analysis of E. coli Lysate on Proteomix® SAX-NP3, Demonstration of High Loading Capacity
HPLC Analysis of E. coli Lysate on Proteomix® SAX-NP3, Demonstration of High Loading Capacity application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SAX-NP3 Strong Anion Exchange Column
L × I.D. 5 cm × 4.6 mm, 3 μm particle size
CONDITIONS
column
Proteomix SAX-NP3, 5 cm x 4.6mm I.D., 3 μm particles (Z777219)
mobile phase
(A) 20 mM Tris, pH 9.0; (B) A + 0.5 M NaCl
gradient
0 - 100% B in 30 min
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
as indicated in figure
sample
2.5 mg/mL
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies