HPLC Analysis of a Peptide Drug on Antibodix® WCX-NP10, Gradient Optimization to Separate Impurity
![HPLC Analysis of a Peptide Drug on Antibodix® WCX-NP10, Gradient Optimization to Separate Impurity application for HPLC](/deepweb/assets/sigmaaldrich/product/images/133/487/a577f420-83f4-4bce-8c12-83282d391c7e/800/a577f420-83f4-4bce-8c12-83282d391c7e.jpg)
CONDITIONS
column
Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase
[A] 3 mM potassium phosphate, pH 5.0; [B] 20 mM potassium phosphate, pH 7.5
gradient
(A) 0% - 50% B (20 min); (B) 0% - 20% B (20 min), 20% B after 20 min
flow rate
1 mL/min
column temp.
ambient
detector
injection
5 μL
sample
2 mg/mL, proprietary peptide drug, pI 7-8
Description
Legal Information
Antibodix is a registered trademark of Sepax Technologies