HPLC Analysis of Barbiturates on Discovery® C18
![HPLC Analysis of Barbiturates on Discovery® C18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/278/941/d7f1daac-e518-49f2-8dce-532a5049b861/800/d7f1daac-e518-49f2-8dce-532a5049b861.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] acetonitrile; [B] 25 mM potassium phosphate buffer, pH 6.9 (25:75, A:B)
flow rate
1.5 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
25 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany