Merck
GC Analysis of 1-Methoxy-2-Propylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX B-PH
GC Analysis of 1-Methoxy-2-Propylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX™ B-PH suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-PH Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
1-Methoxy-2-propylamine
95%
(S)-1-Methoxy-2-propylamine
ChiPros®, produced by BASF, 99%
CONDITIONS
column
Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 μm (71023AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
1 μL, 80:1 split
sample
3 mg/mL in methanol
peaks 1 & 2: 1-methoxy-2-propylamine enantiomers (N-acetyl derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC