US EPA Method 8270: GC Analysis of Semivolatiles on Equity-5
CONDITIONS
column
Equity-5, 30 m × 0.25 mm I.D., 0.5 μm (28092-U)
oven
40 °C (2 min), 50 °C/min to 100 °C, 10 °C/min to 200 °C, 30 °C/min to 325 °C (7.5 min)
inj. temp.
280 °C
detector
MSD, scan range 45-450 amu, 325 °C
carrier gas
pressure programmed, 20 psi (0.0 min), ramp to 80 psi (0.0 min), ramp to 16.5 psi (3 min),ramp to 25 psi (hold for remainder of run)
injection
1 μL, splitless (0.61 min)
liner
4 mm I.D., single taper
sample
50 ng on-column of a primary analyte list consisting of a 74 component semivolatile standard, 6 internal standards, and 8 surrogates
Description
Legal Information
null