Merck
GC Analysis of C1-C5 Hydrocarbons on Alumina Sulfate PLOT
GC Analysis of C1-C5 Hydrocarbons on Alumina Sulfate PLOT suitable for GC
CONDITIONS
column
Alumina sulfate PLOT, 50 m x 0.53 mm I.D. (28324-U)
oven
35 °C (2.5 min), 5 °C/min to 150 °C (4.5 min)
column temp.
200 °C
detector
FID, 230 °C
carrier gas
helium, 3.0 mL/min
injection
10 μL direct using a sampling valve, 150 °C
sample
18-component C1-C5 hydrocarbon standard in nitrogen, each analyte at 25 ng on-column