Merck
  • A mouse model of MYCN-driven retinoblastoma reveals MYCN-independent tumor reemergence.
A mouse model of MYCN-driven retinoblastoma reveals MYCN-independent tumor reemergence.
The Journal of clinical investigation (2017-02-07)
Nan Wu, Deshui Jia, Breanna Bates, Ryan Basom, Charles G Eberhart, David MacPherson
ABSTRACT
The most frequent focal alterations in human retinoblastoma are mutations in the tumor-suppressor gene retinoblastoma (RB) and amplification of the oncogene MYCN. Whether MYCN overexpression drives retinoblastoma has not been assessed in model systems. Here, we have shown that Rb inactivation collaborates strongly with MYCN overexpression and leads to retinoblastoma in mice. Overexpression of human MYCN in the context of Rb inactivation increased the expression of MYC-, E2F-, and ribosome-related gene sets, promoted excessive proliferation, and led to retinoblastoma with anaplastic changes. We then modeled responses to MYCN-directed therapy by suppressing MYCN expression in MYCN-driven retinoblastomas. Initially, MYCN suppression led to proliferation arrest and partial tumor regression with loss of anaplasia. However, over time, retinoblastomas reemerged, typically without reactivation of human MYCN or amplification of murine Mycn. A subset of returning retinoblastomas showed genomic amplification of a Mycn target gene encoding the miR cluster miR-17~92, while most retinoblastomas reemerged without clear genetic alterations in either Mycn or known Mycn targets. This Rb/MYCN model recapitulates key genetic driver alterations seen in human retinoblastoma and reveals the emergence of MYCN independence in an initially MYCN-driven tumor.
Sigma-Aldrich
Monoclonal Anti-p107 antibody produced in mouse, clone KAB6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit