Merck
  • MPI depletion enhances O-GlcNAcylation of p53 and suppresses the Warburg effect.
MPI depletion enhances O-GlcNAcylation of p53 and suppresses the Warburg effect.
eLife (2017-06-24)
Nataly Shtraizent, Charles DeRossi, Shikha Nayar, Ravi Sachidanandam, Liora S Katz, Adam Prince, Anna P Koh, Adam Vincek, Yoav Hadas, Yujin Hoshida, Donald K Scott, Efrat Eliyahu, Hudson H Freeze, Kirsten C Sadler, Jaime Chu
ABSTRACT
Rapid cellular proliferation in early development and cancer depends on glucose metabolism to fuel macromolecule biosynthesis. Metabolic enzymes are presumed regulators of this glycolysis-driven metabolic program, known as the Warburg effect; however, few have been identified. We uncover a previously unappreciated role for Mannose phosphate isomerase (MPI) as a metabolic enzyme required to maintain Warburg metabolism in zebrafish embryos and in both primary and malignant mammalian cells. The functional consequences of MPI loss are striking: glycolysis is blocked and cells die. These phenotypes are caused by induction of p53 and accumulation of the glycolytic intermediate fructose 6-phosphate, leading to engagement of the hexosamine biosynthetic pathway (HBP), increased O-GlcNAcylation, and p53 stabilization. Inhibiting the HBP through genetic and chemical methods reverses p53 stabilization and rescues the Mpi-deficient phenotype. This work provides mechanistic evidence by which MPI loss induces p53, and identifies MPI as a novel regulator of p53 and Warburg metabolism.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-OGT, affinity isolated antibody