Merck
Hypermethylation downregulates P2X7 receptor expression in astrocytoma.
Oncology letters (2018-01-19)
Jing Liu, Ningning Li, Ruofan Sheng, Rui Wang, Zude Xu, Ying Mao, Yin Wang, Ying Liu
ABSTRACT
The present study investigated the altered expression of p2X purinoceptor (P2X7R) in astrocytoma. Reverse transcription-quantitative polymerase chain reaction and western blot analysis were used to determine the P2X7R expression in glioblastoma (GBM) and surrounding normal brain tissue. DNA methylation levels of P2X7R gene promoter in GBM were analyzed using a Sequenom MassARRAY® System. Immunohistochemistry (IHC) was used to detect the expression of P2X7R in astrocytoma at different malignancy grades, including diffuse astrocytoma, anaplastic astrocytoma and GBM. P2X7R mRNA and protein were significantly decreased in GBM compared with normal brain tissues. IHC results showed a negative correlation between P2X7R expression and tumor grade. The decreased P2X7R expression was mostly attributed to hypermethylation of its promoter. Therefore, P2X7R was found to perform an important role in tumorigenesis and progression of astrocytoma.
Sigma-Aldrich
Monoclonal Anti-p53−Biotin antibody produced in mouse, ~1 mg/mL, clone DO-1, ascites fluid, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid