Merck
  • Fungus-derived hydroxyl radicals kill hepatic cells by enhancing nuclear transglutaminase.
Fungus-derived hydroxyl radicals kill hepatic cells by enhancing nuclear transglutaminase.
Scientific reports (2017-07-08)
Ronak Shrestha, Rajan Shrestha, Xian-Yang Qin, Ting-Fang Kuo, Yugo Oshima, Shun Iwatani, Ryutaro Teraoka, Keisuke Fujii, Mitsuko Hara, Mengqian Li, Azusa Takahashi-Nakaguchi, Hiroji Chibana, Jun Lu, Muyi Cai, Susumu Kajiwara, Soichi Kojima
ABSTRACT
We previously reported the importance of induced nuclear transglutaminase (TG) 2 activity, which results in hepatic cell death, in ethanol-induced liver injury. Here, we show that co-incubation of either human hepatic cells or mouse primary hepatocytes derived from wild-type but not TG2