Merck
  • Caffeine and glucosamine mobility shifts by adduction with 2-butanol depended on interaction energy, charge delocalization, and steric hindrance in ion mobility spectrometry.
Caffeine and glucosamine mobility shifts by adduction with 2-butanol depended on interaction energy, charge delocalization, and steric hindrance in ion mobility spectrometry.
Journal of mass spectrometry : JMS (2017-09-07)
Dairo Meza-Morelos, Roberto Fernandez-Maestre
ABSTRACT
Ion mobility spectrometry (IMS) is an analytical technique that separates gas-phase ions drifting under an electric field according to their size to charge ratio. We used electrospray ionization-drift tube IMS coupled to quadrupole mass spectrometry to measure the mobilities of glucosamine (GH
Sigma-Aldrich
2,6-Di-tert-butylpyridine, ≥97%