Merck
  • Inflammation and beyond: new directions and emerging drugs for treating atherosclerosis.
Inflammation and beyond: new directions and emerging drugs for treating atherosclerosis.
Expert opinion on emerging drugs (2016-12-09)
Marie-Jeanne Bertrand, Jean-Claude Tardif
ABSTRACT
Cardiovascular (CV) atherosclerotic disease remains the leading cause of morbidity and mortality worldwide, despite the advances in contemporary therapies. Inflammation is an important process in atherosclerosis, leading to plaque rupture and acute coronary syndrome. Although statin therapy has substantially reduced CV events in primary and secondary prevention, many treated patients will have recurrent adverse CV events despite the standard of care. Thus, drug development aiming to target inflammatory pathways seems a promising avenue for novel therapies in atherosclerosis. Areas covered: Statins have been extensively studied and are the most effective lipid-lowering drugs available for CV prevention. Novel anti-inflammatory drugs are being tested in Phase II and III trials, targeting pathways like interleukin-1, leukotrienes, TNF-α, P-selectin, CCL2-CCR2 and MAP Kinase. Expert opinion: Novel anti-inflammatory therapies seem promising additions to address the residual CV risk present despite the current standard of care, but large clinical trials have not yet shown beneficial effects on clinical events. PCSK9 inhibitors have been shown to substantially reduce LDL-C, however their long-term safety and effects on CV risk are currently being investigated. Pharmacogenomics holds great potential in future lipid trials, enabling the identification of patients who will respond with greater benefits and smaller risk to therapies and to decrease failure rates in drug development, as genotype-dependent effects of the CETP inhibitor dalcetrapib were shown in the dal-OUTCOMES and dal-PLAQUE-2 trials.
Sigma-Aldrich
Dalcetrapib, ≥97% (HPLC)