Merck
  • Developing antineoplastic agents that target peroxisomal enzymes: cytisine-linked isoflavonoids as inhibitors of hydroxysteroid 17-beta-dehydrogenase-4 (HSD17B4).
Developing antineoplastic agents that target peroxisomal enzymes: cytisine-linked isoflavonoids as inhibitors of hydroxysteroid 17-beta-dehydrogenase-4 (HSD17B4).
Organic & biomolecular chemistry (2017-09-05)
Mykhaylo S Frasinyuk, Wen Zhang, Przemyslaw Wyrebek, Tianxin Yu, Xuehe Xu, Vitaliy M Sviripa, Svitlana P Bondarenko, Yanqi Xie, Huy X Ngo, Andrew J Morris, James L Mohler, Michael V Fiandalo, David S Watt, Chunming Liu
ABSTRACT
Cytisine-linked isoflavonoids (CLIFs) inhibited PC-3 prostate and LS174T colon cancer cell proliferation by inhibiting a peroxisomal bifunctional enzyme. A pull-down assay using a biologically active, biotin-modified CLIF identified the target of these agents as the bifunctional peroxisomal enzyme, hydroxysteroid 17β-dehydrogenase-4 (HSD17B4). Additional studies with truncated versions of HSD17B4 established that CLIFs specifically bind the C-terminus of HSD17B4 and selectively inhibited the enoyl CoA hydratase but not the d-3-hydroxyacyl CoA dehydrogenase activity. HSD17B4 was overexpressed in prostate and colon cancer tissues, knocking down HSD17B4 inhibited cancer cell proliferation, suggesting that HSD17B4 is a potential biomarker and drug target and that CLIFs are potential probes or therapeutic agents for these cancers.
Sigma-Aldrich
MISSION® esiRNA, targeting human HSD17B4
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture