Merck
  • Augmented pH-sensitivity absorbance of a ruthenium(ii) bis(bipyridine) complex with elongation of the conjugated ligands: an experimental and theoretical investigation.
Augmented pH-sensitivity absorbance of a ruthenium(ii) bis(bipyridine) complex with elongation of the conjugated ligands: an experimental and theoretical investigation.
Physical chemistry chemical physics : PCCP (2017-09-16)
Siew San Tan, Susumu Yanagisawa, Kouji Inagaki, Yoshitada Morikawa, Mohammad B Kassim
ABSTRACT
An absorbance-based sensor employing ruthenium bipyridyl with a phenanthroline-fused benzoylthiourea moiety formulated as [Ru(ii)(bpy)
Sigma-Aldrich
1,10-Phenanthrolin-5-amine, 97%