Merck
  • Down-regulation of MBD4 contributes to hypomethylation and overexpression of CD70 in CD4
Down-regulation of MBD4 contributes to hypomethylation and overexpression of CD70 in CD4
Clinical epigenetics (2017-10-12)
Wei Liao, Mengying Li, Haijing Wu, Sujie Jia, Nu Zhang, Yong Dai, Ming Zhao, Qianjin Lu
ABSTRACT
Systemic lupus erythematosus (SLE) is an autoimmune disease that is characterized by lymphocytic infiltration and overproduction of autoantibodies, leading to significant morbidity and mortality. However, the pathogenesis of this disorder has not yet been completely elucidated. It has been reported that CD70, a B cell costimulatory molecule encoded by the gene Our results showed that the expression of MBD4 was significantly decreased in CD4 Our results indicated that downregulation of MBD4 contributed to overexpression and hypomethylation of the CD70 gene in SLE CD4
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
MISSION® esiRNA, targeting human MBD4