- X-shaped benzoylbenzophenone derivatives with crossed donors and acceptors for highly efficient thermally activated delayed fluorescence.
X-shaped benzoylbenzophenone derivatives with crossed donors and acceptors for highly efficient thermally activated delayed fluorescence.
Dalton transactions (Cambridge, England : 2003) (2015-01-20)
Sae Youn Lee, Takuma Yasuda, In Seob Park, Chihaya Adachi
PMID25599337
ABSTRACT
Thermally activated delayed fluorescence (TADF) materials based on benzoylbenzophenone, AcPmBPX and PxPmBPX, were designed and synthesized. Organic light-emitting diodes using these materials as emitters exhibited high external electroluminescence quantum efficiencies of up to 11%.
MATERIALS