Merck
Reactivity of the parent amido complexes of iridium with olefins: C-NH
Dalton transactions (Cambridge, England : 2003) (2017-08-19)
Inmaculada Mena, Pilar García-Orduña, Víctor Polo, Fernando J Lahoz, Miguel A Casado, Luis A Oro
ABSTRACT
Herein we report on the different chemical reactivity displayed by two mononuclear terminal amido compounds depending on the nature of the coordinated diene. Hence, treatment of amido-bridged iridium complexes [{Ir(μ-NH
Sigma-Aldrich
1,4-Bis(diphenylphosphino)butane, 98%
Sigma-Aldrich
1,3-Bis(diphenylphosphino)propane, 97%