Merck
  • Effect of advanced oxidation on N-nitrosodimethylamine (NDMA) formation and microbial ecology during pilot-scale biological activated carbon filtration.
Effect of advanced oxidation on N-nitrosodimethylamine (NDMA) formation and microbial ecology during pilot-scale biological activated carbon filtration.
Water research (2017-02-19)
Dong Li, Ben Stanford, Eric Dickenson, Wendell O Khunjar, Carissa L Homme, Erik J Rosenfeldt, Jonathan O Sharp
ABSTRACT
Water treatment combining advanced oxidative processes with subsequent exposure to biological activated carbon (BAC) holds promise for the attenuation of recalcitrant pollutants. Here we contrast oxidation and subsequent biofiltration of treated wastewater effluent employing either ozone or UV/H
Sigma-Aldrich
2-(Trimethylsilyl)ethanethiol, 95%