- Low levels of extracellular glucose limit cardiac anaerobic metabolism in some species of fish.
Low levels of extracellular glucose limit cardiac anaerobic metabolism in some species of fish.
The Journal of experimental biology (2017-06-10)
Kathy A Clow, Connie E Short, William R Driedzic
PMID28596211
ABSTRACT
There is a wide interspecific range in plasma glucose levels in teleosts from less than 0.5 to greater than 10 mmol l
MATERIALS