Merck
SO
Chemistry, an Asian journal (2017-07-13)
Wan-Yin Fang, Jing Leng, Hua-Li Qin
ABSTRACT
A one-pot Pd-catalyzed carbonylation of phenols into their corresponding aryl carboxylic acids and esters through the insertion of carbon monoxide has been developed. This procedure offers a direct synthesis of aryl carboxylic acids and esters from inexpensive and abundant starting materials (phenols, SO
Sigma-Aldrich
4-Cyanobenzoic acid, 99%