Merck
  • Characterization of a novel murine model for spontaneous hemorrhagic stroke using in vivo PET and MR multiparametric imaging.
Characterization of a novel murine model for spontaneous hemorrhagic stroke using in vivo PET and MR multiparametric imaging.
NeuroImage (2017-05-06)
Salvador Castaneda Vega, Christine Weinl, Carsten Calaminus, Lisa Wang, Maren Harant, Walter Ehrlichmann, Dennis Thiele, Ursula Kohlhofer, Gerald Reischl, Johann-Martin Hempel, Ulrike Ernemann, Leticia Quintanilla Martinez, Alfred Nordheim, Bernd J Pichler
ABSTRACT
The clinical use of Magnetic Resonance Imaging (MRI) and Positron Emission Tomography (PET) has proven to be a strong diagnostic tool in the field of neurology. The reliability of these methods to confirm clinical diagnoses has guided preclinical research to utilize these techniques for the characterization of animal disease models. Previously, we demonstrated that an endothelial cell-specific ablation of the murine Serum Response Factor (Srf
Sigma-Aldrich
Mannose triflate, For PET imaging, ≥98% (TLC)