Merck
  • Feasibility of urinary extracellular vesicle proteome profiling using a robust and simple, clinically applicable isolation method.
Feasibility of urinary extracellular vesicle proteome profiling using a robust and simple, clinically applicable isolation method.
Journal of extracellular vesicles (2017-07-19)
Irene V Bijnsdorp, Olga Maxouri, Aarzo Kardar, Tim Schelfhorst, Sander R Piersma, Thang V Pham, Andre Vis, R Jeroen van Moorselaar, Connie R Jimenez
ABSTRACT
Extracellular vesicles (EVs) secreted by prostate cancer (PCa) cells contain specific biomarkers and can be isolated from urine. Collection of urine is not invasive, and therefore urinary EVs represent a liquid biopsy for diagnostic and prognostic testing for PCa. In this study, we optimised urinary EV isolation using a method based on heat shock proteins and compared it to gold-standard ultracentrifugation. The urinary EV isolation protocol using the Vn96-peptide is easier, time convenient (≈1.5 h) and no special equipment is needed, in contrast to ultracentrifugation protocol (>3.5 h), making this protocol clinically feasible. We compared the isolated vesicles of both ultracentrifugation and Vn96-peptide by proteome profiling using mass spectrometry-based proteomics (
Sigma-Aldrich
Anti-TSG101 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution