Merck
One-Pot Hybrid SnO
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-04-04)
Gianvito Caputo, Alice Scarpellini, Francisco Palazon, Athanassia Athanassiou, Despina Fragouli
ABSTRACT
The localized in situ formation of tin dioxide (SnO