Merck
  • Rapid quantitation of avenanthramides in oat-containing products by high-performance liquid chromatography coupled with triple quadrupole mass spectrometry (HPLC-TQMS).
Rapid quantitation of avenanthramides in oat-containing products by high-performance liquid chromatography coupled with triple quadrupole mass spectrometry (HPLC-TQMS).
Food chemistry (2017-02-06)
Zhuohong Xie, Tracy Mui, Marsha Sintara, Boxin Ou, Jodee Johnson, YiFang Chu, Marianne O'shea, Prabhakar Kasturi, Yumin Chen
ABSTRACT
Avenanthramides (AVNs) are a family of nitrogen-containing phenolic compounds produced in oat; AVN 2c, 2p, and 2f are the three major members. An LC-MS/MS method was developed, with the limit of detection (LOD) and the limit of quantitation (LOQ) being, respectively, 0.29ng/mL and 1.96ng/mL for AVN 2c, 0.24ng/mL and 0.60ng/mL for AVE 2p, and 0.42ng/mL and 2.2ng/mL for AVN 2f. The method was validated in oat-containing hot cereal and snack bar samples. The recovery of AVN 2c, 2p, and 2f from these two oat products was 95-113%, and the relative standard deviations ranged from 5% to 9%. This method was used to evaluate oat products and raw oat samples. The effects of location and variety on AVN composition were investigated. The method presented here provides a novel and rapid tool to quantitate the abundance of AVN 2c, 2p, and 2f in oat-containing products.
Product Description
Supelco
Avenanthramide B, analytical standard