Merck
  • High-throughput untargeted screening of veterinary drug residues and metabolites in tilapia using high resolution orbitrap mass spectrometry.
High-throughput untargeted screening of veterinary drug residues and metabolites in tilapia using high resolution orbitrap mass spectrometry.
Analytica chimica acta (2017-01-22)
Wei Jia, Xiaogang Chu, James Chang, Perry G Wang, Ying Chen, Feng Zhang
ABSTRACT
An analytical method was developed and validated for simultaneous analysis of one hundred and thirty-seven veterinary drug residues and metabolites from sixteen different classes in tilapia utilizing an improved fully non-targeted way of data acquisition with fragmentation. The automated on-line extraction procedure was achieved in a simple disposable pipet extraction. Ultrahigh-performance liquid chromatography and electrospray ionization quadrupole Orbitrap high-resolution mass spectrometry (UHPLC Q-Orbitrap) was used for the separation and detection of all the analytes. The methodology was validated by taking into consideration the guidelines specified in European SANCO/12571/2013 Guideline 2013 and Commission Decision 2002/657/EC. The extraction recoveries ranged from 81% to 111%. The limits of decision ranged from 0.01 to 2.73 μg kg
Sigma-Aldrich
3-Aminocoumarin, 97%
Supelco
AMOZ-d5, VETRANAL®, analytical standard