Merck
  • Enhancing analysis throughput, sensitivity and specificity in LC/ESI-MS/MS assay of plasma 25-hydroxyvitamin D
Enhancing analysis throughput, sensitivity and specificity in LC/ESI-MS/MS assay of plasma 25-hydroxyvitamin D
Journal of pharmaceutical and biomedical analysis (2017-01-13)
Shoujiro Ogawa, Hiroki Kittaka, Akiho Nakata, Kenji Komatsu, Takahiro Sugiura, Mamoru Satoh, Fumio Nomura, Tatsuya Higashi
ABSTRACT
The plasma/serum concentration of 25-hydroxyvitamin D
Sigma-Aldrich
4-(Methylamino)benzoic acid, 97%