Merck
  • Thyronamines and Derivatives: Physiological Relevance, Pharmacological Actions, and Future Research Directions.
Thyronamines and Derivatives: Physiological Relevance, Pharmacological Actions, and Future Research Directions.
Thyroid : official journal of the American Thyroid Association (2016-09-22)
Carolin Stephanie Hoefig, Riccardo Zucchi, Josef Köhrle
ABSTRACT
Thyronamines (3-T
Sigma-Aldrich
T1AM, ≥98% (HPLC)