Merck
  • Synthesis and in vitro antioxidant and antimicrobial studies of novel structured phosphatidylcholines with phenolic acids.
Synthesis and in vitro antioxidant and antimicrobial studies of novel structured phosphatidylcholines with phenolic acids.
Food chemistry (2016-12-17)
Marrapu Balakrishna, Shiva Shanker Kaki, Mallampalli S L Karuna, Sripada Sarada, C Ganesh Kumar, R B N Prasad
ABSTRACT
Novel phenoylated phosphatidylcholines were synthesized from 1,2-dipalmitoyl phosphatidylcholine/egg 1,2-diacyl phosphatidylcholine and phenolic acids such as ferulic, sinapic, vanillic and syringic acids. The structures of phenoylated phosphatidylcholines were confirmed by spectral analysis. 2-acyl-1-lyso phosphatidylcholine was synthesized from phosphatidylcholine via regioselective enzymatic hydrolysis and was reacted with hydroxyl protected phenolic acids to produce corresponding phenoylated phosphatidylcholines in 48-56% yields. Deprotection of protected phenoylated phosphatidylcholines resulted in phenoylated phosphatidylcholines in 87-94% yields. The prepared compounds were evaluated for their preliminary in vitro antimicrobial and antioxidant activities. Among the active derivatives, compound 1-(4-hydroxy-3,5-dimethoxy) cinnamoyl-2-acyl-sn-glycero-3-phosphocholine exhibited excellent antioxidant activity with EC50 value of 16.43μg/mL. Compounds 1-(4-hydroxy-3-methoxy) cinnamoyl-2-acyl-sn-glycero-3-phosphocholine and 1-(4-hydroxy-3,5-dimethoxy) cinnamoyl-2-palmitoyl-sn-glycero-3-phosphocholine exhibited good antioxidant activity with EC50 values of 36.05 and 33.35μg/mL respectively. Compound 1-(4-hydroxy-3-methoxy) cinnamoyl-2-palmitoyl-sn-glycero-3-phosphocholine exhibited good antibacterial activity against Klebsiella planticola with MIC of 15.6μg/mL and compound 1-(4-hydroxy-3-methoxy) benzoyl-2-acyl-sn-glycero-3-phosphocholine exhibited good antifungal activity against Candida albicans with MIC of 15.6μg/mL.
Product Description
Supelco
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard